Imprimer
VWR®, Centrifuge Tubes, Soda-Lime Glass
Fournisseur: VWR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VWR®
525-1575EA 66.2 EUR
525-1575 525-1574 525-1573 525-1572 525-1571 525-1570 525-1577 525-1576
VWR®, Centrifuge Tubes, Soda-Lime Glass
Tubes Tubes à centrifuger
Available with round or conical bottom, graduated or non graduated. Not suitable for applications related with strong heat or rapid temperature changes.
  • Produced from soda-lime glass tubing
  • Withstand forces up to 3000 ×g
  • Graduations in high contrast blue enamel
Ideal for use as disposables. Can be autoclaved but the temperature difference (ΔT) should not exceed 40 °C.