Imprimer
Hydroxy naphthol blue disodium salt ACS, Sigma-Aldrich®
Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Attention
Synonyme(s): Hydroxynaphthol blue sodium salt, Hydroxy napthol blue, Sodium (E)-3-hydroxy-4-((2-hydroxy-6-sulpho-3-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1-sulphonate
M-Clarity™ (Niveau de Qualité) = MQ200
219916-100G 219916-5G
219916-100GEA 118 EUR
219916-100G 219916-5G
Hydroxy naphthol blue disodium salt ACS, Sigma-Aldrich®
Hydroxy naphthol blue disodium salt
Hydroxy naphthol blue disodium salt, ACS reagent.
Hydroxy naphthol blue disodium salt is a chemical compound belonging to the class of azo dyes commonly used as a pH indicator.It exists as small blue crystals. Hydroxy naphthol blue is suitable for the quantification of metal leaching in metal affinity chromatography. It changes color at pH range 12 to 13 from blue to reddish pink, in the presence of calcium ions and to deep blue in the presence of disodium ethylenediaminetetraacetic acid (EDTA).
Formule: C₂₀H₁₂N₂O₁₁S₃·2Na
Poids moléculaire: 598.479 g/mol
Point de fusion: 274 °C
Densité: 2,130 g/cm³
Température de stockage: Room Temperature
Numéro MDL: MFCD00004075
Numéro CAS: 165660-27-5
Commander maintenant
Résultats des tests de spécification
Appearance (Form) Powder or Crystals or Granules or Chunks
Suitability for Calcium Determination Pass
Meets ACS Requirements Current ACS Specification
Recommended Retest Period 3 Years