Imprimer
RNaseOUT™ RNAse removal solution
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-70 786-71
786-70EA 110 EUR
786-70 786-71
RNaseOUT™ RNAse removal solution
Produits nettoyants Décontaminants
RNaseOUT™ is uniquely formulated to destroy and remove RNases on contact.
  • Simply spray and rinse
  • RNaseOUT™ is non-toxic and residue free
  • RNaseOUT™ allows common equipment to be safely and confidently used for all RNA work