Imprimer
Hand cleansers, hydro alcoholic gel
Fournisseur: Basan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2602359 2602367 2602458
BSNC2602359EA 127 EUR
BSNC2602359 BSNC2602367 BSNC2602458
Hand cleansers, hydro alcoholic gel
Soin de la peau Savons et soins nettoyants
Ready-to-use hydro-alcoholic gel, for fast and total hand disinfection. Extra soft for the skin.
  • Usable in all sectors
  • Without colouring, fragrance-free, non allergenic
  • Colourless; alcohol odour
  • Fungicidal, bactericidal, and virucidal properties, and active against the Bird Flu virus
  • Non-greasy product when applied
  • Packaging 100% recyclable
Wash and dry your hands before use. Take a dose of hydro alcoholic gel, rub your hands for a min and 15 seconds, allow to dry.
Made of ethanol: 74,25%, isopropanol: 6,5%. 1 L of this gel allows 550 doses.
Certifications: Food contact approved. Product (CE 648/2004). EN 1040 - Bactericidal test.
Attention: Use biocides safely. Always read the label and product information before use. Protect against freezing temperatures.