Imprimer
Hand disinfectants, Tork
Fournisseur: Essity
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
511103 420103 590103
SCAA511103EA 103 EUR
SCAA511103 SCAA420103 SCAA590103
Hand disinfectants, Tork
Soin de la peau Désinfectants Mousses et liquides désinfectants
These hand disinfectants contains 80% denatured alcohol (w/w) and does not need to be rinsed. This is developed for regular use without sticky residues. Ideal for healthcare and even meets the requirements for scrub rooms.
  • Disinfects the hands quickly
  • Prevent infections with the highly effective
  • Effective disinfectant gel
The gel is odourless and features moisturizing ingredients that protect the hands.
Attention: Use biocides safely. Always read the label and product information before use.