Imprimer
Surface disinfectant, Gammaclean 1512B
Fournisseur: Basan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2502023 2502044 2502074
BSNC2502023EA 0 EUR
BSNC2502023 BSNC2502044 BSNC2502074
Surface disinfectant, Gammaclean 1512B
Produits nettoyants Désinfectants
Gammaclean 1512B is a powerful disinfectant degreaser that cleans and disinfects all surfaces in the food industry (tables, stainless steel utensils, floors and walls) and hospitals in one single step.
Certifications: Meets EN1040, EN1276, EN13697, AFNOR T72-201, EN14476 standards.
Attention: Use biocides safely. Always read the label and product information before use.