Imprimer
Bag clips for Stomacher® bags
Fournisseur: Seward (FermionX)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BA6099 BA6097
432-3116EA 566 EUR
432-3116 SEWABA6097
Bag clips for Stomacher® bags
Portoirs Porte-sachets
Specifically designed for Seward Stomacher® 400 bags.
  • Eliminates any leakage during storage or incubation
  • Provides a reusable alternative to closure bags
  • Easily cleaned with most lab disinfectants (not autoclavable)
These bag clips allow fast and easy securing of the Stomacher® bags.