Imprimer
Immersion oil viscosity 1,250 cSt (lit.), Sigma-Aldrich®
Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
I0890-100ML I0890-500ML
I0890-100MLEA 65.1 EUR
I0890-100ML I0890-500ML
Immersion oil viscosity 1,250 cSt (lit.), Sigma-Aldrich®
Stains Microscopy and staining reagents
Immersion oil has been used in epifluorescent microscopy and cell identification.
  • Suitable for Hematology
  • Suitable for Histology
Immersion oil has a larger impact on 3D resolution. Specifically, matching the immersion oil refractive index to glass, reliably enhances the ability to visualize dendritic spines.