Imprimer
Rapid yeast and mold count plates, Petrifilm™
Marques : 3M Food Safety
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6475 6477
MMMP6475EA 144 EUR
MMMP6475 MMMP6477
Rapid yeast and mold count plates, Petrifilm™
Milieux pour microbiologie Milieux prêts à l'emploi de microbiologie
3M™ Petrifilm™ Rapid Yeast and Mold Count Plates simplify and accelerate detection. Featuring a new indicator technology that makes colonies easier to interpret, these completely sample-ready plates return results in just 48 hours.
  • Utilizes the convenience and efficiency of the 3M™ Petrifilm™ Plate platform
  • Proven cost effective tool for the detection of yeast and mold
  • Results in 48 hours
  • Proven as reliable as DRBC and DG-18 spread plate methods, as well as acidified PDA and YGC agar pour plate methods
  • Can be used with both low and high water activity foods
  • Eliminates the need for the labor intensive, multiple plate spread plate technique
  • Versatility for air, swab or surface contact environmental sampling
Yeasts and molds can grow in a wide range of conditions on your raw materials, products and process environment, making them difficult to control. Complicating matters is the five-day incubation period required by traditional agar methods. However, 3M™ Petrifilm™ Rapid Yeast and Mold Count Plates are ready in just 48 hours.