Imprimer
Rapid coliform count plates, Petrifilm™
Marques : 3M Food Safety
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6402 6412
710-0781EA 97 EUR
710-0781 710-0771
Rapid coliform count plates, Petrifilm™
Milieux pour microbiologie Milieux prêts à l'emploi de microbiologie
3M™ Petrifilm™ Rapid coliform count plates are designed to provide fast, accurate results in detecting coliform contamination.
  • Utilises the convenience and efficiency of the 3M™ Petrifilm™ plate platform
  • Proven cost effective tool for the detection of coliform
  • Results in 4 to 24 hours
  • This plate has been awarded Official Methods of Analysis (OMA) #200015 for Food and NF VALIDATION certificates: 3M-01/5-03/97 A for all Human food products , 3M-01/5-03/97 B for All humand food products and 3M-01/5-03/97 C for All humand food products (except process pork products) by AFNOR Certification
  • Food processors and contract labs worldwide trust 3M™ Petrifilm™ plates to provide accurate, efficient, consistent results
High coliform contamination can be detected beginning at 4 hours with distinctive yellow zones. Presumptive coliforms may be be recorded at 14 hours. Confirmed results are available at 24 hours.