Imprimer
Compact system for 250 ml extraction, behrotest® KEX 250 / KEX 250 F
Fournisseur: BEHR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B00217732 B00217737
BEHRB00217732EA 1830 EUR
BEHRB00217732 BEHRB00217737
Compact system for 250 ml extraction, behrotest® KEX 250 / KEX 250 F
Chambres anaérobies
behrotest® serial extraction devices are the economical and user-friendly solution for classic Soxhlet and fat extraction. With behr hydrolysis units (six sample positions), acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
  • Complete with reaction vessels, extraction spacers and coolers
  • Power is individually adjustable for each sample position
  • Cooling water distribution ensures even cooling at all sample positions
  • Practical stand for holding condensers securely between extractions
  • Holders for secure fixation of extraction adapters after removal of the sample vessels
Extractors with specially developed lifting tube (Böckerhoff type) guarantee homogeneous results at all sample locations
S models: The use of extractors with a stopcock does away with the need for any additional distillation after extraction.
W×D×H (in cm): 23×33×80
Controller: No
Power consumption (in W): 450
Current Consumption (in A): 3
Sample positions: 1
Capacity (in ml): 500
Flask size (in ml): 500
Extractor size (in ml): 250