Imprimer
RackScan
Fournisseur: EPPENDORF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6002000.006 6002000.014
EPPE6002000.006EA 11450 EUR
EPPE6002000.006 EPPE6002000.014
RackScan
Systèmes de code-barres
The RackScan instrument can read up to 96 barcoded vials in parallel.
  • Camera based instrument resulting in rapid imaging and decoding
  • Comfortable and easy plug-in system
  • Small footprint, ideal for benchtop (and integration)
The RackScan can be equipped with the 's' accessory.