Imprimer
Trichrome stain kit (Masson) light green variation
Marques : RAL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
361350-0000
RALA361350-0000EA 288 EUR
RALA361350-0000
Trichrome stain kit (Masson) light green variation
Trichrome stain
Staining for Histological and Cytological studies of Fixed and Paraffined Tissue Sections
Staining with kit Trichrome de Masson, light green variation enables a trichromic station on histological sections with a specific coloration of the collagen by Light Green.
Mayer Haemalum stains nuclei black-blue or brown. Ponceau Fuchsin solution stains cytoplams red pink. Light green stains collagen green.
Components - Name (ref) Packing
MAYER HAEMALUM (Ref 320550) 100 ml (Others packagings 1000, 2500 ml)
FUCHSIN PONCEAU (Ref 361770) 100 ml (Others packagings 1000, 2500 ml)
1% ACETIC WATER (Ref 361840) 200 ml
ACID PHOSPHOMOLYBDIQUE (Ref 300140) 100 ml
IVDLogo