Imprimer
VWR® Adhesive Sealing Films for PCR and Storage
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
391-0622EA 75.3 EUR
391-0622 391-0643
VWR® Adhesive Sealing Films for PCR and Storage
Seals Microplate Seals
Polyester seals feature a strong adhesive to minimize evaporation during PCR and storage applications.
  • Ideal for PCR & storage
  • Certified DNase- and RNase- free
  • Temperature range is –40 to +120 °C
  • Non-pierceable
  • Minimizes evaporation
Removes easily without leaving adhesive residue.