Imprimer
VWR® Nitrile Light Ocean Blue, Nitrile Gloves
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
112-6070EA 22.6 EUR
112-6070 112-6067 112-6069
VWR® Nitrile Light Ocean Blue, Nitrile Gloves
Gants
These light, ambidextrous, durable gloves are ideal for food handling, catering and kitchen work, lab applications, and other tasks with complex risks.
  • Nonsterile, powder-free
  • Low thickness and textured fingertips for excellent grip
  • Particularly skin friendly for users with latex allergy
  • Rolled cuffs minimise the risk of tearing
  • Level 2 – AQL 1,5
These thin nitrile gloves are free from phthalates, softeners, and allergenic latex proteins.
Certifications: EN ISO 374-1 Type B (KTP), EN ISO 374-5 VIRUS; Cat. III; AQL 1,5
Food contact approved.