Imprimer
Core samplers
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5310-0001 5310-0002 5310-0003
331-0374EA 664 EUR
331-0374 331-0375 331-0376
Core samplers
Samplers Solid Samplers
These core samplers can be used to take samples of bulk materials, which can easily become stuck or settle during sampling due to their particle form.
  • Stainless steel AISI 316
  • For bulk materials that become stuck or settle easily
In comparison to standard all-layer samplers for bulk materials, the two-part core sampler has no gap dimensions in which the medium can become stuck while it is being removed. This makes it possible to take a proper sample of powders and granulates through multiple layers, such as glass balls or metal powders.
Depending on the size, the stainless steel sampler can be used for a wide range of different containers, for example big bags or small containers. The bulk material sampler takes a representative core sample and is ideal for use in various laboratory and industrial applications.
Informations de livraison: Delivery includes core sampler (consisting of boring rod and boring tube) and fixing clip.