Imprimer
ThermoSafe® FreezeTherm® Pre-Qualified Dry Ice Shippers
Fournisseur: Sonoco Thermosafe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FT58 FT103 FT6 FT7 FT14 FT17 FT29 FT54 FT269 FT396 FT176 FT44
SCATFT58EA 608 EUR
SCATFT58 SCATFT103 SCATFT6 SCATFT7 SCATFT14 SCATFT17 SCATFT29 SCATFT54 SCATFT269 SCATFT396 SCATFT176 SCATFT44
ThermoSafe® FreezeTherm® Pre-Qualified Dry Ice Shippers
Boîtes d'expédition et de transport
FreezeTherm® dry ice shippers are pre-qualified to several external ambient temperatures to give users confidence of likely performance.
  • High-quality EPS base and lid inside a strong, outer corrugated box
  • Outer box has all necessary dry ice markings to simplify the shipment
FreezeTherm® dry ice shipping systems provide ready access to convenient, affordable protection for frozen products in transit. The wide range of off-the-shelf sizes means there’s a FreezeTherm® solution for every application.