Imprimer
Heating circulators, MAGIO™ MS and MX series
Fournisseur: JULABO GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9032504 9033506 9033512 9033526
461-1423EA 4900 EUR
461-1423 461-1424 461-1425 461-1426
Heating circulators, MAGIO™ MS and MX series
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
The MAGIO™ series of heating circulators operate with a working temperature range of 20 to +300 °C and heating capacity of up to 3 kW. With their powerful pressure/suction pump the MAGIO™ units are ideal for external temperature applications. An extensive range of accessories and excellent dynamics mean that the MAGIO™ circulators can be modularly and individually adapted to applications.
  • Integrated programmer allows temperature time profiles to be run
  • Stainless steel wetted parts mean units are ideal for demanding external applications
  • Automatic shut off in the event of high temperature or low liquid level to avoid any potential damage
  • Bright, easy to read display
  • Bath tanks have high quality thermal insulation to save energy and a built in drain tap
  • Powerful, continuously adjustable pressure/suction pump
The high resolution, modern colour TFT touch display has a multi-lingual user interface and shows all important information at a glance. Three large, predefined main screens clearly display data and graphics with various application priorities. All display functions are easy to operate with a fingertip.
Modern interfaces make remote control easy and practical data management possible as well as allowing integration into process structures. All MAGIO™ units have integrated PT100 connection, USB interface, RS232 and Ethernet. Analogue interfaces are available as optional accessories.
Max. flow rate / max. pressure:
31 L/min, 0,92 bar (pressure), 0,4 bar (suction)
Certifications: Classification III according to DIN 12876-1
IP protection class according to DIN EN 60529: IP 20.