Imprimer
Hook2Loop Bungee tool tether, DBI-SALA®, 3M™
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DBI-SALA®
MMMA1500047EA 33.6 EUR
MMMA1500047 CPSD1500048
Hook2Loop Bungee tool tether, DBI-SALA®, 3M™
Protections anti-chutes Câbles de sécurité
Bungee tool tether with self-locking carabiner.
  • Adjusts to virtually any wrist size with hook and loop system
  • Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
  • Elastic band helps ensure a comfortable fit
  • Attached trigger can be used to quickly work with many hand tools
This is a very versatile extension that can be used with virtually any tool, such as spud wrenches, scaffold wrenches, adjustable wrenches, and more.
Designed for use in general industrial, oil and gas, mining, construction, and transportation industries.