Imprimer
Halt™ Phosphatase Inhibitor Cocktail
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
78420 78426 78427 78428
PIER78420EA 237 EUR
PIER78420 PIER78426 PIER78427 PIER78428
Halt™ Phosphatase Inhibitor Cocktail
Suppléments et additifs pour culture cellulaire
Phosphorylation and dephosphorylation is a molecular on/off switch that regulates a number of key biological pathways within the cell, including signal transduction, cell division, and apoptosis. The Halt Phosphatase Inhibitor Cocktail preserves phosphorylation of proteins in cell lysates and tissue extracts.
  • All in one format - Halt Phosphatase Inhibitor Cocktail inhibits both serine/threonine as well as protein tyrosine phosphatases
  • Compatible with standard protein assays
  • User friendly storage at 2 to 8 °C - Halt Phosphatase Inhibitor Cocktail does not require frozen storage
  • Convenient size - 1 ml of cocktail protects up to 100 ml of sample
The cocktail contains a mixture of four phosphatase inhibitors of broad specificity, including sodium fluoride, sodium orthovanadate, sodium pyrophosphate and -glycerophosphate. Unlike other commercially available phosphatase inhibitor cocktails, which protect against either serine/threonine phosphatases or protein tyrosine phosphatases, the Halt Phosphatase Inhibitor Cocktail protects phosphoproteins from both families of phosphatases. Conveniently packaged in a 100X format, the cocktail is stable at 4 °C and is compatible with common protein assays, including Bradford-based assays.