Imprimer
Adjustable wristbands, DBI-SALA®
Fournisseur: CAPITAL SAFETY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1500085 1500087
CPSD1500085EA 0 EUR
CPSD1500085 CPSD1500087
Adjustable wristbands, DBI-SALA®
Dispositifs de protection antistatique Bracelets antistatiques
Adjustable wristbands are available with retractor and elastic strap with hook and loop, or with cord and elastic strap with hook and loop.
  • Adjusts to virtually any wrist size with hook and loop system
  • Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
  • Elastic band helps ensure a comfortable fit
  • Attached trigger can be used to quickly work with many hand tools
The DBI-SALA® Adjustable wristband with retractor is constructed out of elastic webbing that can be adjusted to virtually any wrist size. Tools can easily be tied off to this wristband using sewn in cord.
Wristbands are designed for use in general industrial, oil and gas, mining, construction, and transportation industries.