Imprimer
Safety funnels with lid, 'MARCO' V2.0
Fournisseur: S.C.A.T.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
318951 318955 318950 318954 318952 318953 318990 318991 318992 318993 318994 318995
221-0315EA 107 EUR
221-0315 SCTV318955 SCTV318950 SCTV318954 SCTV318952 SCTV318953 SCTV318990 SCTV318991 SCTV318992 SCTV318993 SCTV318994 SCTV318995
Safety funnels with lid, 'MARCO' V2.0
Capuchons Bouchons de sécurité
S.C.A.T. 'MARCO' V2.0 safety funnels can be closed safely and prevent danger of ignition.
  • 'MARCO' V2.0 safety lid funnel is a modern lid funnel made of high quality HDPE with extremely high resistance to acids, alkalis and many chemicals
  • Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents explosive or health-endangering gases escaping
  • Sturdy hinge guarantees the closing mechanism has a long service life
  • Rotating screw cap makes it easier to screw the lid funnel on and off the canister
  • Removable sieve for easy cleaning
  • The non conductive version of the 'MARCO' V2.0 safety lid funnel is in line with the SCAT colour scheme with its blue and white colouring, the conductive version is black, this difference in colour helps differentiate the conductive from the non conductive version
The innovative 'MARCO' V2.0 safety funnel is a high quality lid funnel for the disposal of liquid waste. The lid, with integrated rubber lip, seals the funnel and prevents the escape of toxic or explosive vapours. Numerous features guarantee safe handling. Available in different thread sizes for safe attachment to laboratory bottles, canisters or other containers.
Diameter: 140 mm