Imprimer
Handheld automated analytical syringe, eVol® XR
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2910020 2910021 2910022 2910023 2910024 2910025 2910026 2910027 2910320 2910322 2910324 2910029 2910028 2910329 2910035 2910037 2910335 2910205
548-1349EA 105 EUR
548-1349 548-1350 548-1351 548-1352 548-1353 548-1354 548-1427 548-1428 548-1431 548-1432 548-1433 548-1430 548-1429 548-1434 548-1515 548-2355 548-2356 548-1514
Handheld automated analytical syringe, eVol® XR
Syringes Syringes for Chromatography
eVol® is the coupling of two precision devices: A digitally controlled electronic drive and an XCHANGE® enabled analytical syringe. The result is a digitally controlled positive displacement dispensing system that can be programmed to reproducibly and accurately perform a wide variety of liquid handling procedures.
  • Precise user-independent handling procedures
  • XCHANGE® analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids and methods preventing possible cross-contamination of reagents
  • Perfect solution for accurately aspirating and dispensing both aqueous and non aqueous liquids
Typical applications: Preparation of calibration standards, addition of internal standards, precise dispensing of non aqueous liquids and sample dilution. Only five XCHANGE® syringes are required to dispense liquid volumes covering the range from 0,2 to 1000 μl.
eVol® features a familiar Touch Wheel user interface and a full-colour screen. The smart menu allows all the functions to be accessed logically and quickly. The programming functions are intuitive and include help screens and prompts.
To comply with stringent laboratory standards (e.g. GLP, GMP, FDA) eVol® can be easily calibrated to ensure accurate dispensing at all times. The gravimetric calibration procedure is simple, intuitive and can be performed at appropriate intervals. Calibration factors can be stored for up to ten XCHANGE® syringes and can be quickly loaded when the syringe is changed.
Note: All SGE 25 to 250 µl replacement needles can be used with the 50 to 100 µl eVol® syringes. All SGE 1 to 2,5 ml replacement needles can be used with the 500 µl eVol® syringe.
Informations de livraison: eVol® XR automated analytical syringe starter kit is supplied with eVol® XR electronic syringe, three eVol® syringes; 5 and 100 µl and 1 ml, stand, universal charger, instruction manual and disc with manual in multiple languages. eVol® NMR starter kit contains eVol® electronic syringe (handle), 3 eVol® syringes - 5 µl syringe is supplied with a 115 mm needle, 50 and 500 µl syringes are supplied with both 115 and 180 mm needles, stand, universal charger, comprehensive instruction manual, disc with manual in multiple languages.