Imprimer
Electroporation solution kits, BTXpress™
Marques : BTX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
45-0805 45-0802
732-1285EA 867 EUR
732-1285 732-1284
Electroporation solution kits, BTXpress™
Nucleic Acid Reagents Transformation Reagents
The BTXpress™ is a single buffer solution, developed to quickly and efficiently deliver genes into mammalian cells that were previously considered “hard to transfect” by chemical and other non viral methods. This solution, in combination with the BTX electroporation instruments, provides researchers with the versatility and success needed across a broad range of cell types. Transfection using this high performance electroproation solution is equally effective in delivering DNA as well as siRNA into mammalian cells.
  • High efficiency transfection of cell lines and previously considered hard to transfect cells
  • Low toxicity resulting in improved cell viability
  • One buffer used in place of standard electroporation buffers for all mammalian cell types
  • Versatility of choosing electroporation optimisation settings for the highest transfection efficiency with BTX instruments
  • Increased numbers of tranfections per kit providing better value
As a universal solution the BTXpress™ electroporation reagent can be used with electroporation generators, including Amaxa™ Nucleofector®.
Informations de livraison: BTXpress™ high performance electroporation solution is supplied either as a kit, which includes the BTX Plus cuvettes with transfer pipettes, or as a buffer alone.