Imprimer
Serological pipettes, Standard Line
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-3707EA 202 EUR
612-3707 612-3705 612-3704 612-3703 612-3702 612-3701 612-3700 612-3699 612-3698 612-3697 612-3696 612-3695
Serological pipettes, Standard Line
Pipettes Serological Pipettes
100% high-clarity polystyrene, graduated, sterile.
  • Graduations are calibrated for accurate dispensing to within ±3%
  • Colour-coded stripes for quick volume identification
  • Non pyrogenic
  • With cotton plug
Cdt: Individually wrapped in paper-plastic bags or bulk packed in bags.