Imprimer
ESD safety shoes, lace-up, uvex 1 sport hygiene, S3, 6582
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6582.1/35 6582.1/36 6582.1/37 6582.1/38 6582.1/39 6582.1/40 6582.1/41 6582.1/42 6582.1/43 6582.1/44 6582.1/45 6582.1/46 6582.1/47 6582.1/48 6582.1/49 6582.1/50 6582.1/51 6582.1/52 6582.2/35 6582.2/36 6582.2/37 6582.2/38 6582.2/39 6582.2/40 6582.2/41 6582.2/42 6582.2/43 6582.2/44 6582.2/45 6582.2/46 6582.2/47 6582.2/48 6582.2/49 6582.2/50 6582.2/51 6582.2/52 6582.3/35 6582.3/36 6582.3/37 6582.3/38 6582.3/39 6582.3/40 6582.3/41 6582.3/42 6582.3/43 6582.3/44 6582.3/45 6582.3/46 6582.3/47 6582.3/48 6582.3/49 6582.3/50 6582.3/51 6582.3/52
114-8904EA 103 EUR
114-8904 114-8905 114-8906 114-8907 114-8908 114-8909 114-8910 114-8911 114-8912 114-8913 114-8914 114-8915 114-8916 114-8917 114-8918 114-8919 114-8920 114-8921 114-8922 114-8923 114-8924 114-8925 114-8926 114-8927 114-8928 114-8929 114-8930 114-8931 114-8932 114-8933 114-8934 114-8935 114-8936 114-8937 114-8938 114-8939 114-8940 114-8941 114-8942 114-8943 114-8944 114-8945 114-8946 114-8947 114-8948 114-8949 114-8950 114-8951 114-8952 114-8953 114-8954 114-8955 114-8956 114-8957
ESD safety shoes, lace-up, uvex 1 sport hygiene, S3, 6582
Chaussures et bottes
Modern, metal-free, ultra-lightweight and flexible S3 safety lace-up shoe.
  • Upper made from water repellent microfibre material that is easy to clean
  • Minimal number of seams to reduce contamination
  • Suitable for people allergic to chrome as it’s made from synthetic materials
  • Washable at 30 °C
  • All sole materials are free of silicones, plasticisers, and other paint wetting impairment substances
  • Meets the ESD specification with volume resistance less than 35 megaohms
  • Soft padding on collar and tongue
100% metal-free uvex xenova® toe cap, with a compact design, anatomical shape, good lateral stability, and no thermal conductivity. The ergonomically designed outsole is comprised of dual-density polyurethane with good shock absorption and excellent slip resistance.
A removable anti-static comfortable insole provides the foot with exceptional, full surface shock absorption ensuring comfort in high strain zones.
Certifications: EN ISO 20345:2011 S3 SRC.