Imprimer
General purpose gloves, A109
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A109ORRL A109ORRXL A109ORRM
PRTWA109ORRLEA 27.5 EUR
PRTWA109ORRL PRTWA109ORRXL PRTWA109ORRM
General purpose gloves, A109
Gants
The lines of gloves for general use and gloves with grip are the most popular. A careful selection of coverings, included latex, nitrile, polyurethane and PVC, makes our gloves suitable for many use.
  • Materials: 65% polyester, 35% cotton, latex
  • 100% breathable seamless lining
  • Tested for both cut and heat protection
  • Smooth PU coating for increased abrasion resistance
  • Palm dipped for greater dexterity and ventilation
A wide range of protective gloves is available. They are used manufactured fom the best materials.
Certifications: AS/NZS 2161 .3; AS/NZS 2161 .2; EN 388:2016 +A1:2018 - (3.1.4.3.X); EN 420 certified.