Imprimer
Bump caps, vent cool, PW69
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PW69BKR PW69GRR PW69NAR PW69RER
PRTWPW69BKREA 18 EUR
PRTWPW69BKR PRTWPW69GRR PRTWPW69NAR PRTWPW69RER
Bump caps, vent cool, PW69
Casques Casquettes
Sport look bump cap, with large mesh panel on the top, allowing maximum airflow, reflective stripes for enhanced safety and style.
  • Long peak (70 mm) for maximum protection against shock and light
  • Inner ABS shell and soft pad for shock absorption
  • Durable high density EVA protects and gives comfort to the wearer
  • Retail bag which aids presentation for retail sales
  • Hook and loop with adjustable size
Materials: cotton, mesh, ABS, EVA.
Certifications: EN 812