Imprimer
Safety helmets, Endurance, PS53
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PS53RBR PS53BKR PS53YER PS53WHR PS53ORR
PRTWPS53RBREA 28.2 EUR
PRTWPS53RBR PRTWPS53BKR PRTWPS53YER PRTWPS53WHR PRTWPS53ORR
Safety helmets, Endurance, PS53
Casques Casques de sécurité
Designed to be specially used for working at heights. Light, comfortable and compact with a short peak. ABS shell, textile comfort harness six points, wheel ratchet adjustment, size 52 to 63 cm.
  • Light and robust
  • Specially designed for working at height
  • Electrical insulation up to 1000Vac or 1500Vdc (EN 50365)
  • Six points textile harness
  • Wheel ratchet size adjustment for easy fitting
  • Hi-Visibility colour for visibility in daylight
Comfortable and lightweight, the PPE range will ensure a pleasant wear, even for long periods of use.
Certifications: ANSI/ISEA Z89.1 TYPE I Class E; EN 50365 Class:0; EN 397 -30°C/+50°C
Informations de livraison: Improved technical sweat band and four points chin strap (Y style) with soft rubber chin protector included.