Imprimer
High visibility waistcoat, C494
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C494YER4X/5X C494ORRS/M C494ORRL/XL C494YERL/XL C494ORRXX/3X C494YERS/M C494ORR4X/5X C494YERXX/3X
PRTWC494YER4X/5XEA 6.3 EUR
PRTWC494YER4X/5X PRTWC494ORRS/M PRTWC494ORRL/XL PRTWC494YERL/XL PRTWC494ORRXX/3X PRTWC494YERS/M PRTWC494ORR4X/5X PRTWC494YERXX/3X
High visibility waistcoat, C494
Tenue de travail
This vest made from 100% polyester, warp knit features two bands of reflective tape on the body.
  • Hook and loop closure for easy access
  • Generous fit for wearer comfort
  • Certified to EN ISO 20471 after 50× washes
Cooling mesh upper torso for increased breathability. Dual ID holder allows use of both portrait and landscape ID card display.
Attention: Machine wash warm, mild process. Do not bleach. Line dry. Do not iron. Do not dry clean.