Imprimer
Anti impact cut resistant gloves, A722
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A722G8RM A722G8RL A722G8RXXXL A722G8RXL A722G8RXXL A722G8RS
PRTWA722G8RMEA 258 EUR
PRTWA722G8RM PRTWA722G8RL PRTWA722G8RXXXL PRTWA722G8RXL PRTWA722G8RXXL PRTWA722G8RS
Anti impact cut resistant gloves, A722
Gants
Anti impact cut resistant gloves combines superior cut protection and maximum impact protection using TPR protective pods across the back of the hand.
  • ANSI cut resistance level A4
  • Reinforced thumb crotch
  • Padded palms
  • Hook and loop strap for safe secure fit
  • HPPE, glass fibre and polyester liner material
  • 100% breathable seamless liner
Palms are dipped in a nitrile sandy coating, providing exceptional grip in water, grease or oil contact.
Certifications: Standards: AS/NZS 2161 .3. AS/NZS 2161 .2. ANSI/ISEA 105 - 2016 CUT Level (A4). EN 388:2003 - 4.5.4.3. EN 388:2016 - 2.1.4.2.X. CE marked. EN 420.