Imprimer
Flame resistant trousers, BizFlame™ Plus, FR26
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FR26NARXXXL FR26NATM FR26NATXXL FR26NARXL FR26NARXXL FR26NARL FR26NARM FR26NARS FR26NAR4XL FR26NATXL FR26NARXS FR26NATL
PRTWFR26NARXXXLEA 75.5 EUR
PRTWFR26NARXXXL PRTWFR26NATM PRTWFR26NATXXL PRTWFR26NARXL PRTWFR26NARXXL PRTWFR26NARL PRTWFR26NARM PRTWFR26NARS PRTWFR26NAR4XL PRTWFR26NATXL PRTWFR26NARXS PRTWFR26NATL
Flame resistant trousers, BizFlame™ Plus, FR26
Tenue de travail
This anti-static trouser is constructed with high visibility reflective tape double stitched for enhanced durability. Features include triple stitched seams, high visibility strips on legs, front fly brass zip and adjustable bottom leg opening.
  • Guaranteed flame resistance for life of garment
  • Certified protection against molten metal splash
  • Sew on flame resistant industrial wash tape
  • 50+ UPF rated fabric to block 98% of UV rays
  • Durable, strong and long lasting brass zip
  • Half elasticated waist for a secure and comfortable fit
Two tier knee pad pockets allowing two positioning options, ruler pocket. Protection against radiant, convective and contact heat.
Shell fabric : BizFlame™ Plus 350 g
Certifications: Standards: ELIM TESTING IN PROGRESS, EN 1149 -5, IEC 61482-2 IEC 61482-1-2 Class 1, EN ISO 11612 (A1+A2, B1, C1, E2, F1), EN ISO 11611 Class 2 (A1+A2), ASTM F1959/F1959M-12 (ATPV=13.6 Cal/CM2, (HAF=82%))