Imprimer
Climbing helmets, PW97
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PW97WHR PW97RBR PW97YER
PRTWPW97WHREA 17.6 EUR
PRTWPW97WHR PRTWPW97RBR PRTWPW97YER
Climbing helmets, PW97
Casques Casques de sécurité
Lightweight and ventilated helmet for workers at height. High wear comfort and secure fit through ergonomically formed shell. The six point ratchet adjustable harness uses nylon suspension for wearer comfort.
  • Y chin strap
  • Molten metal
Certifications: EN 397 (–30/+50°C MM 440V), ANSI/ISEA Z89.1 Type I.