Imprimer
Flame retardant high visibility multi overalls, Araflame, AF91
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AF91ORRL AF91ORRM AF91ORRS AF91ORRXL AF91ORRXXXL AF91ORRXXL AF91ORR4XL AF91ORR5XL
PRTWAF91ORRLEA 346 EUR
PRTWAF91ORRL PRTWAF91ORRM PRTWAF91ORRS PRTWAF91ORRXL PRTWAF91ORRXXXL PRTWAF91ORRXXL PRTWAF91ORR4XL PRTWAF91ORR5XL
Flame retardant high visibility multi overalls, Araflame, AF91
Tenue de travail
This anti-static overall twinned with the silver flame resistant reflective tape on the shoulders, torso, arms and legs provides superior enhanced visibility.
  • Class 1 welding protection
  • Flame resistant industrial wash tape
  • Durable, strong and long lasting brass zip
  • Radio loop for easy clipping of a radio
  • Six pockets for ample storage
  • 50+ UPF rated fabric to block 98% of UV rays
  • Side access pockets
Inherent flame resistant qualities will not diminish with washing. Protection against radiant, convective and contact heat.
Weight: 260 g
Certifications: EN ISO 11612 (A1, B1, C1, F1), EN 1149 -5, EN 13034 Type 6 (fabric only), RIS-3279-TOM ISSUE 1, GO/RT 3279 Issue 8, EN ISO 11611 Class 1 (A1), EN ISO 20471 Class 3.