Imprimer
Infrared rapid digestion devices (manually adjustable power setting) for nitrogen determination according to Kjeldahl, behrotest® InKjel
Fournisseur: BEHR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B00218067 B00218068 B00218101 B00218103 B00373235
705-0432EA 5200 EUR
705-0432 705-0433 705-0430 705-0431 BEHRB00373235
Infrared rapid digestion devices (manually adjustable power setting) for nitrogen determination according to Kjeldahl, behrotest® InKjel
Systèmes de digestion Systèmes de minéralisation
behrotest® InKjel digestion units are equipped with a high-performance infrared heater.
  • Direct heating of the samples by the infrared heater prevents the endlessly long heating times required using conventional heating block systems
  • High quality quartz heaters instead of the common steel tube heater ensures particularly homogeneous heating in all sample positions
Quality and position of the behr infrared heater guarantee the user identical heating phases and digestion temperatures in all sample positions. This also applies to the double-row arrangement of racks for 12 samples. The glass digestion vessels are suspended in the rack and are not deposited at the bottom of the digestion unit, therefore the digestion units are less susceptible to breakage than in an aluminium heating block.
This makes the behrotest® InKjel the ideal digestion system for nitrogen determination according to Kjeldahl and other high temperature digestions. In Kjeldahl digestions, the temperature is adjusted through the boiling point of sulphuric acid.