Imprimer
Avantor® Hichrom, Retention Gap/Precolumn, DPTMDS Deactivated High Temperature Fused Silica
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HI98-05-DHT-1EA 24.6 EUR
HI98-05-DHT-1 HI98-05-DHT-2 HI98-05-DHT-5 HI98-05-DHT-10 HI98-10-DHT-1 HI98-10-DHT-2 HI98-10-DHT-5 HI98-10-DHT-10 HI98-10-DHT-20 HI98-15-DHT-1 HI98-15-DHT-2 HI98-15-DHT-5 HI98-15-DHT-10 HI98-15-DHT-20 HI98-15-DHT-30 HI98-18-DHT-1 HI98-18-DHT-2 HI98-18-DHT-5 HI98-18-DHT-10 HI98-18-DHT-20 HI98-18-DHT-30 HI98-20-DHT-1 HI98-20-DHT-2 HI98-20-DHT-5 HI98-20-DHT-10 HI98-20-DHT-20 HI98-20-DHT-30 HI98-25-DHT-1 HI98-25-DHT-2 HI98-25-DHT-5 HI98-25-DHT-10 HI98-25-DHT-20 HI98-25-DHT-30 HI98-25-DHT-60 HI98-32-DHT-1 HI98-32-DHT-2 HI98-32-DHT-5 HI98-32-DHT-10 HI98-32-DHT-20 HI98-32-DHT-30 HI98-32-DHT-60 HI98-53-DHT-1 HI98-53-DHT-2 HI98-53-DHT-5 HI98-53-DHT-10 HI98-53-DHT-20 HI98-53-DHT-30 HI98-53-DHT-60
Avantor® Hichrom, Retention Gap/Precolumn, DPTMDS Deactivated High Temperature Fused Silica
Colonnes pour chromatographie
DPTMDS deactivated high temperature fused silica retention gap / precolumn. General purpose deactivation for both apolar and polar solvents.