Imprimer
Avantor® Hichrom, Retention Gap/Precolumn, CW Deactivated Fused Silica
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HI97-05-D-1EA 22.4 EUR
HI97-05-D-1 HI97-05-D-2 HI97-05-D-5 HI97-05-D-10 HI97-10-D-1 HI97-10-D-2 HI97-10-D-5 HI97-10-D-10 HI97-10-D-20 HI97-15-D-1 HI97-15-D-2 HI97-15-D-5 HI97-15-D-10 HI97-15-D-20 HI97-15-D-30 HI97-18-D-1 HI97-18-D-2 HI97-18-D-5 HI97-18-D-10 HI97-18-D-20 HI97-18-D-30 HI97-20-D-1 HI97-20-D-2 HI97-20-D-5 HI97-20-D-10 HI97-20-D-20 HI97-20-D-30 HI97-25-D-1 HI97-25-D-2 HI97-25-D-5 HI97-25-D-10 HI97-25-D-20 HI97-25-D-30 HI97-25-D-60 HI97-32-D-1 HI97-32-D-2 HI97-32-D-5 HI97-32-D-10 HI97-32-D-20 HI97-32-D-30 HI97-32-D-60 HI97-53-D-1 HI97-53-D-2 HI97-53-D-5 HI97-53-D-10 HI97-53-D-20 HI97-53-D-30 HI97-53-D-60
Avantor® Hichrom, Retention Gap/Precolumn, CW Deactivated Fused Silica
Colonnes pour chromatographie
CW deactivated fused silica retention gap / precolumn. Deactivation mainly for polar solvents.