Imprimer
Avantor® Hichrom, Retention Gap/Precolumn, HMDS Deactivated Fused Silica
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
HI96-05-D-1EA 22.4 EUR
HI96-05-D-1 HI96-05-D-2 HI96-05-D-5 HI96-05-D-10 HI96-10-D-1 HI96-10-D-2 HI96-10-D-5 HI96-10-D-10 HI96-10-D-20 HI96-15-D-1 HI96-15-D-2 HI96-15-D-5 HI96-15-D-10 HI96-15-D-20 HI96-15-D-30 HI96-18-D-1 HI96-18-D-2 HI96-18-D-5 HI96-18-D-10 HI96-18-D-20 HI96-18-D-30 HI96-20-D-1 HI96-20-D-2 HI96-20-D-5 HI96-20-D-10 HI96-20-D-20 HI96-20-D-30 HI96-25-D-1 HI96-25-D-2 HI96-25-D-5 HI96-25-D-10 HI96-25-D-20 HI96-25-D-30 HI96-25-D-60 HI96-32-D-1 HI96-32-D-2 HI96-32-D-5 HI96-32-D-10 HI96-32-D-20 HI96-32-D-30 HI96-32-D-60 HI96-53-D-1 HI96-53-D-2 HI96-53-D-5 HI96-53-D-10 HI96-53-D-20 HI96-53-D-30 HI96-53-D-60
Avantor® Hichrom, Retention Gap/Precolumn, HMDS Deactivated Fused Silica
Colonnes pour chromatographie
HMDS deactivated fused silica retention gap / precolumn. Deactivation mainly for apolar solvents.