Imprimer
VWR® Hitachi ChromAssist, data station
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
906-0123EA 2090 EUR
906-0123 906-0124 HITA903-0374 HITA903-0375
VWR® Hitachi ChromAssist, data station
Software Chromatography Software
The newly developed ChromAssist Data Station combines the proven intuitive structure of the Hitachi software family with an innovative new design. As a chromatography data system (CDS), ChromAssist contains all the necessary functions for controlling VWR® Hitachi HPLC and UHPLC instruments, including method creation, sample table, data integration and reporting.
  • New user-friendly design
  • Integrated training videos and help functions
  • Simple creation of methods, sample tables
  • Innovative report editor
  • Advanced GLP functionality
Additionally, some indicators have been added to give the user a good overview of his analysis and the instrument at any time. ChromAssist software also includes useful GLP tools that meet the requirements of a modern laboratory and simplify the documentation of important instrument parameters.
As a special highlight, the software includes a comprehensive range of training videos and help texts that make it easy for new HPLC users to get started and can also help experienced users with "troubleshooting" or instrument maintenance.
Supported devices from the VWR® HITACHI LC family:
- Chromaster
- Chromaster UltraRs
- LaChrom Elite
- LaChrom Ultra
System requirements:
- Operating System: Microsoft® Windows® 10 Pro
- CPU: Intel® Core i5-6400 processor (6th generation) or later
- RAM: 8 GB or more
- Display: 1920 × 1080 (Full HD) or more
Attention: Does not comply with 21 CFR Part 11, therefore not applicable in strong regulated environments. If you need software with this standard, please search for DataApex ClarityVA or Agilent Openlab in our webshop.