Imprimer
Phosphorus AAS standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E4054-100ML E4054-500ML E4114-100ML
HONCE4054-100MLEA 44.3 EUR
HONCE4054-100ML HONCE4054-500ML HONCE4114-100ML
Phosphorus AAS standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de phosphore
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).