Imprimer
Mercury AAS standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E4033-100ML E4033-500ML
HONCE4033-100MLEA 45 EUR
HONCE4033-100ML HONCE4033-500ML
Mercury AAS standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de mercure
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).