Imprimer
Gallium AAS Standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E4029-100ML E4029-500ML
HONCE4029-100MLEA 112 EUR
HONCE4029-100ML HONCE4029-500ML
Gallium AAS Standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de gallium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).