Imprimer
Beryllium AAS Standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E4009-100ML E4010-100ML E4010-500ML
HONCE4009-100MLEA 192 EUR
HONCE4009-100ML HONCE4010-100ML HONCE4010-500ML
Beryllium AAS Standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de béryllium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).