Imprimer
Chromium(VI) ICP standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E1512-100ML E1513-100ML
HONCE1512-100MLEA 157 EUR
HONCE1512-100ML HONCE1513-100ML
Chromium(VI) ICP standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de chrome
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).