Imprimer
Lithium ICP and ICP-MS standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E1329-100ML E1397-100ML
HONCE1329-100MLEA 126 EUR
HONCE1329-100ML HONCE1397-100ML
Lithium ICP and ICP-MS standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de lithium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).