Imprimer
Chromium ICP & ICP-MS standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E1312-100ML E1380-100ML
HONCE1312-100MLEA 126 EUR
HONCE1312-100ML HONCE1380-100ML
Chromium ICP & ICP-MS standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de chrome
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).