Imprimer
Gallium ICP Standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E1022-100ML E1149-100ML
HONCE1022-100MLEA 132 EUR
HONCE1022-100ML HONCE1149-100ML
Gallium ICP Standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de gallium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).