Imprimer
Europium ICP Standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E1020-100ML E1147-100ML
HONCE1020-100MLEA 146 EUR
HONCE1020-100ML HONCE1147-100ML
Europium ICP Standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards d'europium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).