Imprimer
Barium ICP standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
E1008-100ML E1101-100ML E1135-100ML E1205-100ML
HONCE1008-100MLEA 101 EUR
HONCE1008-100ML HONCE1101-100ML HONCE1135-100ML HONCE1205-100ML
Barium ICP standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de baryum
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).