Imprimer
Die-cut thermal transfer labels, B-7425
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
149261 149262 121031 121008 148675 148678 148671 121000 121001 121002 121006 121007 121013 121017 121018 121019 121020 121021 121028 121029 148670 148672 148673 148674 148676 148677 148679 148680 148681 148682 148683 148684
BRDY149261EA 162 EUR
BRDY149261 BRDY149262 BRDY121031 BRDY121008 BRDY148675 BRDY148678 BRDY148671 BRDY121000 BRDY121001 BRDY121002 BRDY121006 BRDY121007 BRDY121013 BRDY121017 BRDY121018 BRDY121019 BRDY121020 BRDY121021 BRDY121028 BRDY121029 BRDY148670 BRDY148672 BRDY148673 BRDY148674 BRDY148676 BRDY148677 BRDY148679 BRDY148680 BRDY148681 BRDY148682 BRDY148683 BRDY148684
Die-cut thermal transfer labels, B-7425
Étiquettes
Die-cut thermal transfer labels are made from polypropylene, with a permanent acrylic adhesive and matte finish.
  • General and industrial identification
  • Laboratory identification
  • Temperature identification
  • Die-cut labels are supplied on a roll
  • Maximum service temperature: 70 °C
  • Minimum service temperature: –80 °C
For freezer or liquid nitrogen applications, B-7425 must be applied prior to being frozen.